EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Efficient iodine catalyzed three components domino reaction for the synthesis of 1-((phenylthio)(phenyl)methyl)pyrrolidin-2-one derivatives possessing anticancer activities.
PMID 22695979
Abstract
A simple and efficient three components domino reaction of γ-butyrolactam (2-pyrrolidinone), aromatic aldehyde and substituted thiophenol catalyzed by elemental iodine resulted in the formation of 1-((phenylthio)(phenyl)methyl)pyrrolidin-2-one derivatives. The stability of the synthesized analogues was evaluated in stimulated gastric fluid (SGF) and bovine serum albumin (BSA). In vitro anticancer activity was investigated in the low micromolar range and a few analogues were found to possess good activity. This current protocol provides several advantages like shorter reaction time, excellent yield and convenient work-up.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
83300
2-Pyrrolidinone, purum, ≥98.0% (GC)
C4H7NO