EMAIL THIS PAGE TO A FRIEND
Current radiopharmaceuticals
Maturation of a key resource - the germanium-68/gallium-68 generator: development and new insights.
PMID 22697481
Abstract
(68)Ge/(68)Ga radionuclide generators have been investigated for almost fifty years, since the cyclotron-independent availability of positron emitting (68)Ga via the (68)Ge/(68)Ga system had always attracted researches working in basic nuclear chemistry as well as radiopharmaceutical chemistry. However, it took decades and generations of research (and researchers) to finally reach a level of (68)Ge/(68)Ga radionuclide generator designs adequate to the modern requirements of radiometal labelling chemistry. Nevertheless, most of the existing commercial generator systems address aspects of (68)Ge breakthrough and safe synthesis of (68)Ga radiopharmaceuticals by adopting eluate post-processing technologies. Among the strategies to purify (68)Ga eluates, the cation exchange based version is relevant in terms of purification efficiency. In addition, it offers more options towards further developments of (68)Ga radiopharmaceuticals. Today, one may expect that the (68)Ge/(68)Ga radionuclide generator systems could contribute to the clinical impact of nuclear medicine diagnoses for PET similar to the established (99)Mo/(99m)Tc generator system for SPECT. The exciting perspective for the (68)Ge/(68)Ga radionuclide generator system, in turn, asks for systematic chemical, radiochemical, technological and radiopharmaceutical efforts, to guarantee reliable, highly-efficient and medically approved (68)Ge/(68)Ga generator systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
263230
Germanium, chips, 99.999% trace metals basis
Ge
203343
Germanium, chips, 99.999% trace metals basis
Ge
327395
Germanium, powder, −100 mesh, ≥99.999% trace metals basis
Ge
203351
Germanium, powder, −100 mesh, ≥99.99% trace metals basis
Ge
GF58081451
Germanium, sheet, 50x50mm, thickness 3.0mm, polycrystalline, 99.999%
Ge
GF99720647
Germanium, sheet, 50x50mm, thickness 1.0mm, polycrystalline, 99.999%
Ge
GF31896366
Germanium, sheet, 7x24mm, thickness 1.0mm, polycrystalline, 99.999%
Ge
GF46650090
Germanium, disks, 15mm, thickness 1.0mm, polycrystalline, 100%
Ge
GF90754858
Germanium, disks, 20mm, thickness 1.0mm, single crystal, 100%
Ge
GF77697680
Germanium, microfoil, 25x25mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
Ge
GF58880739
Germanium, microfoil, 25x25mm, thinness 0.5μm, specific density 333μg/cm2, 6 micron aluminum permanent support, 100%
Ge
GF12210624
Germanium, microfoil, 50x50mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
Ge
GF57527219
Germanium, rod, 25mm, diameter 2.0mm, polycrystalline, n-type, 99.999%
Ge
GF65029417
Germanium, rod, 25mm, diameter 5mm, polycrystalline, n-type, 99.999%
Ge
GF14446346
Germanium, rod, 50mm, diameter 5mm, polycrystalline, n-type, 99.999%
Ge
GF04605780
Germanium, rod, 6mm, diameter 6.0mm, single crystal, 100%
Ge
GF09210417
Germanium, sheet, 10x10mm, thickness 0.10mm, polycrystalline, 99.999%
Ge
GF00633664
Germanium, sheet, 10x10mm, thickness 0.25mm, polycrystalline, 99.999%
Ge
GF09629859
Germanium, sheet, 10x10mm, thickness 0.5mm, single crystal, -111, 100%
Ge
GF45727241
Germanium, sheet, 10x10mm, thickness 0.6mm, single crystal, -111, 100%
Ge
GF44621580
Germanium, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Ge
GF44356978
Germanium, sheet, 25x25mm, thickness 1.0mm, single crystal, 99.999%
Ge
GF20558022
Germanium, sheet, 25x25mm, thickness 3.0mm, polycrystalline, 99.999%
Ge
GF49319815
Germanium, sheet, 4x4mm, thickness 0.05mm, polycrystalline, 99.999%
Ge
GF73619107
Germanium, sheet, 50x50mm, thickness 0.5mm, single crystal, 99.999%
Ge
GF37208369
Germanium, sheet, 6x6mm, thickness 1.0mm, polycrystalline, 99.999%
Ge