EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Iron phosphate microflowers as peroxidase mimic and superoxide dismutase mimic for biocatalysis and biosensing.
PMID 22699259
Abstract
Novel iron phosphates microflowers which show SOD-like and peroxidase-like mimic activities were prepared, suggesting potential applications as a biocatalyst and a biosensor for H(2)O(2).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
436011
Iron(III) phosphate dihydrate, Fe 29 %
FeO4P · 2H2O