EMAIL THIS PAGE TO A FRIEND
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology
Spectroscopic studies of the color modulation mechanism of firefly (beetle) bioluminescence with amino-analogs of luciferin and oxyluciferin.
PMID 22699416
Abstract
Spectroscopic properties of amino-analogs of luciferin and oxyluciferin were investigated to confirm the color modulation mechanism of firefly (beetle) bioluminescence. Fluorescence solvatochromic character of aminooxyluciferin analogs indicates that the bioluminescence of aminoluciferin is useful for evaluating the polarity of a luciferase active site.