EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[Hepatoprotective effect of deanol aceglumate on experimental stress-induced gastropathy and diabetes mellitus].
PMID 22702105
Abstract
Experiments on mice with streptozotocin-induced diabetes mellitus and stress-induced erosive ulcerous damage of the mucous membrane of stomach showed evidence of the preventive activity of deanol aceglumate in the course of peroral introduction at a dose of 250 mg/kg per 24 h during 4 days. This effect is accompanied by activation of the peristalsis of bowels and by an increase in the blood flow in the wall of stomach.