EMAIL THIS PAGE TO A FRIEND
Journal of occupational and environmental hygiene
Preliminary studies on the use of acid-soluble cellulose acetate internal capsules for workplace metals sampling and analysis.
PMID 22702231