EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Colorimetric detection of urine glucose based ZnFe2O4 magnetic nanoparticles.
PMID 22702236
Abstract
In this paper, we discovered that ZnFe(2)O(4) magnetic nanoparticles (MNPs) possess intrinsic peroxidase-like activity. ZnFe(2)O(4) MNPs exhibit several advantages such as high catalytic efficiency, good stability, monodispersion, and rapid separation over other peroxidase nanomimetics and horseradish peroxidase (HRP). ZnFe(2)O(4) MNPs were used as a colorimetric biosensor for the detection of urine glucose. This method is simple, inexpensive, highly sensitive, and selective for glucose detection using glucose oxidase (GOx) and ZnFe(2)O(4) MNPs with a linear range from 1.25 × 10(-6) to 1.875 × 10(-5) mol L(-1) with a detection limit of 3.0 × 10(-7) mol L(-1). The color change observable by the naked eyes based on the oxidation of 3,3',5,5'-tetramethylbenzidine (TMB) is the principle for the sensing of urine glucose level.