EMAIL THIS PAGE TO A FRIEND
Journal of pediatric surgery
Childhood thoracic actinomycosis: case report.
PMID 22703819
Abstract
Actinomycosis is a rare bacterial infection. It has a pseudotumoral appearance when localized to the lungs or mediastinum. The diagnosis is often delayed because clinical presentation is nonspecific, bacteriologic diagnosis is difficult, and culture growth is slow. The diagnosis is mainly based upon histologic identification of actinomycotic sulfur granules. Actinomycosis is usually sensitive to penicillin G. Surgical treatment is reserved for failure of medical treatment and complications. We report a case of thoracic actinomycosis in a 13-year-old boy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1000000
Phenoxymethylpenicillin, European Pharmacopoeia (EP) Reference Standard
C16H18N2O5S