EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Absolute quantitative analysis for sorbic acid in processed foods using proton nuclear magnetic resonance spectroscopy.
PMID 22704472
Abstract
An analytical method using solvent extraction and quantitative proton nuclear magnetic resonance (qHNMR) spectroscopy was applied and validated for the absolute quantification of sorbic acid (SA) in processed foods. The proposed method showed good linearity. The recoveries for samples spiked at the maximum usage level specified for food in Japan and at 0.13 g kg(-1) (beverage: 0.013 g kg(-1)) were larger than 80%, whereas those for samples spiked at 0.063 g kg(-1) (beverage: 0.0063 g kg(-1)) were between 56.9 and 83.5%. The limit of quantification was 0.063 g kg(-1) for foods (and 0.0063 g kg(-1) for beverages containing Lactobacillus species). Analysis of the SA content of commercial processed foods revealed quantities equal to or greater than those measured using conventional steam-distillation extraction and high-performance liquid chromatography quantification. The proposed method was rapid, simple, accurate, and precise, and provided International System of Units traceability without the need for authentic analyte standards. It could therefore be used as an alternative to the quantification of SA in processed foods using conventional method.