EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Effect of chronic activation of 5-HT3 receptors on 5-HT3, 5-HT(1A) and 5-HT(2A) receptors functional activity and expression of key genes of the brain serotonin system.
PMID 22705909
Abstract
Among serotonin (5-HT) receptors, the 5-HT(3) receptor is the only ligand-gated ion-channel. Little is known about the interaction between the 5-HT(3) receptor and other 5-HT receptors and influence of 5-HT(3) chronic activation on other 5-HT receptors and the expression of key genes of 5-HT system. Chronic activation of 5-HT(3) receptor with intracerebroventricularly administrated selective agonist 1-(3-chlorophenyl)biguanide hydrochloride (m-CPBG) (14 days, 40 nmol, i.c.v.) produced significant desensitization of 5-HT(3) and 5-HT(1A) receptors. The hypothermic responses produced by acute administration of selective agonist of 5-HT(3) receptor (m-CPBG, 40 nmol, i.c.v.) or selective agonist of 5-HT(1A) receptor (8-hydroxy-2-(di-n-propylamino)tetralin) (8-OH-DPAT, 1mg/kg, i.p.) was significantly lower in m-CPBG treated mice compared with the mice of control groups. Chronic m-CPBG administration failed to induce any significant change in the 5-HT(2A) receptor functional activity and in the expression of the gene encoding 5-HT(2A) receptor. Chronic activation of 5-HT(3) receptor produced no considerable effect on the expression on 5-HT(3), 5-HT(1A), and 5-HT transporter (5-HTT) and tryptophan hydroxylase-2 (TPH-2) genes - the key genes of brain 5-HT system, in the midbrain, frontal cortex and hippocampus. In conclusion, chronic activation of ionotropic 5-HT(3) receptor produced significant desensitization of 5-HT(3) and postsynaptic 5-HT(1A) receptors but caused no considerable changes in the expression of key genes of the brain 5-HT system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C144
1-(3-Chlorophenyl)biguanide hydrochloride, solid
C8H10ClN5 · HCl