EMAIL THIS PAGE TO A FRIEND
Nihon shinkei seishin yakurigaku zasshi = Japanese journal of psychopharmacology
[JSNP Excellent Presentation Award for AsCNP 2011: involvement of 5-HT(1A) receptors in fluvoxamine-induced enhancement of prefrontal dopamine release].
PMID 22708271
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000853
Fluvoxamine for system suitability, European Pharmacopoeia (EP) Reference Standard
C15H21F3N2O2 · C4H4O4
F2802
Fluvoxamine maleate, solid
C15H21F3N2O2 · C4H4O4