EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
A physiologically based pharmacokinetics model for florfenicol in crucian carp and oral-to-intramuscular extrapolation.
PMID 22712485
Abstract
In this study, an oral physiologically based pharmacokinetics (PBPK) model was developed for florfenicol in crucian carp (Carassius auratus). Subsequently, oral-to-intramuscular extrapolation was performed and the two models were used to predict florfenicol concentrations in the edible tissues of crucian carp. The oral model gave good predictions in most tissues, except for kidney and liver in which the florfenicol concentrations were underestimated at the later time points. In contrast, using the intramuscular model, the concentrations in the kidney were overestimated at the later time points. Both models had the best predictive ability in the main edible tissue, the muscle. The oral model also accurately predicted the florfenicol concentrations in the muscle after multiple doses. The present study demonstrated the feasibility of predicting florfenicol concentrations in the edible tissues of crucian carp using a route-to-route extrapolation method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0261
Thiamphenicol
C12H15Cl2NO5S