EMAIL THIS PAGE TO A FRIEND
Geriatrics & gerontology international
Transient ischemic attack caused by delayed orthostatic hypotension in an elderly hypertensive patient.
PMID 22712811
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8277
Midodrine hydrochloride, analytical standard, for drug analysis
C12H18N2O4 · HCl