EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Studies of inhibitor binding to the [4Fe-4S] cluster of quinolinate synthase.
PMID 22715136
Abstract
Stop for NadA! A [4Fe-4S] enzyme, NadA, catalyzes the formation of quinolinic acid in de novo nicotinamide adenine dinucleotide (NAD) biosynthesis. A structural analogue of an intermediate, 4,5-dithiohydroxyphthalic acid (DTHPA), has an in vivo NAD biosynthesis inhibiting activity in E. coli. The inhibitory effect can be explained by the coordination of DTHPA thiolate groups to a unique Fe site of the NadA [4Fe-4S] cluster.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51269
Dihydroxyacetone phosphate hemimagnesium salt hydrate, ≥95% (TLC)
C3H5Mg0.5O6P · xH2O