EMAIL THIS PAGE TO A FRIEND
Chemosphere
Degradation of carbofuran and carbofuran-derivatives in presence of humic substances under basic conditions.
PMID 22717164
Abstract
The influence of humic aggregates in water solution upon the chemical stability of carbofuran (CF) and the carbofuran-derivatives, 3-hydroxy-carbofuran (HCF) and 3-keto-carbofuran (KCF), has been investigated in basic media. An inhibition upon the basic hydrolysis of 3-hydroxy-carbofuran and 3-keto-carbofuran (≈ 1.7 and ≈ 1.5-fold, respectively) was observed and it was rationalized in terms of the micellar pseudophase model. Nevertheless, non-significant effect upon the carbofuran stability was found in the presence of humic substances. These behaviors have been compared with the corresponding ones in other synthetic colloidal aggregates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32056
Carbofuran, PESTANAL®, analytical standard
C12H15NO3
426008
Carbofuran, 98%
C12H15NO3