EMAIL THIS PAGE TO A FRIEND
Journal of cystic fibrosis : official journal of the European Cystic Fibrosis Society
A network meta-analysis of the efficacy of inhaled antibiotics for chronic Pseudomonas infections in cystic fibrosis.
PMID 22717535
Abstract
Various inhaled antibiotics are currently used for treating chronic Pseudomonas aeruginosa lung infection in cystic fibrosis (CF) patients, however their relative efficacies are unclear. We compared the efficacy of the inhaled antibiotics tobramycin (TIP, TIS-T, TIS-B), colistimethate sodium (colistin) and aztreonam lysine for inhalation (AZLI) based on data from randomised controlled trials. In the base case, efficacies of antibiotics were compared using a network meta-analysis of seven trials including change from baseline in forced expiratory volume in 1 second (FEV(1)) % predicted, P. aeruginosa sputum density and acute exacerbations. The tobramycin preparations, AZLI and colistin, showed comparable improvements in efficacy in terms of FEV1% predicted at 4 weeks; the difference in % change from baseline (95%CrI) for TIP was compared to TIS-T (-0.55, -3.5;2.4), TIS-B (-0.64, -7.1;5.7), AZLI (3.64, -1.0;8.3) and colistin (5.77, -1.2;12.8). We conclude that all studied antibiotics have comparable efficacies for the treatment of chronic P. aeruginosa lung infection in CF.