EMAIL THIS PAGE TO A FRIEND
Biopharmaceutics & drug disposition
Pharmacokinetics of dibenzylamine administered in a sustained drug delivery system with cefazolin.
PMID 2271755
Abstract
The pharmacokinetics of dibenzylamine administered in a sustained drug delivery system with cefazolin was studied after i.m. administration of a dose of 1250 mg to healthy volunteers. The serum and urine levels of dibenzylamine were determined by a GLC technique using a specific nitrogen-phosphorus detector. Characterization of the kinetic parameters was performed by applying compartmental and non-compartmental analysis. Dibenzylamine was found to reach concentrations close to 300 ng ml-1 approximately 5 h after administration. The elimination constant had a value of 0.832 +/- 0.821 h-1 (mean +/- S.D.), which is higher than the release constant of the derivative (0.109 +/- 0.072 h-1) (mean +/- S.D.). These results show that release of dibenzylamine may be considered the limiting kinetic process, which governs the elimination of the product from the organism. Only a small amount of dibenzylamine is excreted in urine unchanged 3.43 +/- 3.28 per cent (mean +/- S.D.). Using the pharmacokinetic parameters calculated for dibenzylamine, a prediction has been made of the concentrations reached in a multiple dosage regimen after administration of a dose of 1250 mg every 24 h. The accumulation factor was 1.09.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D34108
Dibenzylamine, 97%
C14H15N