EMAIL THIS PAGE TO A FRIEND
Journal of chromatographic science
Separation and isolation of fusion protein using a new native preparative PAGE device.
PMID 22718746
Abstract
A human serum albumin and Thymosin α1 (HSA-Tα1) fusion protein was designed and over-expressed in Pichia pastoris. To purify the fusion protein, a new native preparative electrophoresis system that involved a modified device with a sample receiving chamber, and an assay method with Coomassie Blue G-250 tracing the collection of the protein of interest. In this device, two gels were run in parallel: native vertical collecting polyacrylamide gel electrophoresis (PAGE) and native vertical tracing PAGE. Samples mixed with or without Coomassie Blue G-250 loading buffer were separately loaded to the two aforementioned gels, and the fractions were collected until the tracing protein band combined with dye reached 1 cm from the sample-receiving chamber at the bottom of the gel. Approximately nine fractions were collected at regular intervals of 15 min. HSA-Tα1 fusion protein with 95% relative homogeneity was harvested and manifested similar immunological activities as synthetic Tα1 after a single-step purification of this preparative PAGE. As a result, this system offers a new, rapid and simple method for the purification of the protein of interest.