EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Switchable hydrophilicity solvents for lipid extraction from microalgae for biofuel production.
PMID 22721685
Abstract
A switchable hydrophilicity solvent (SHS) was studied for its effectiveness at extracting lipids from freeze-dried samples of Botryococcus braunii microalgae. The SHS N,N-dimethylcyclohexylamine extracted up to 22 wt.% crude lipid relative to the freeze-dried cell weight. The solvent was removed from the extract with water saturated with carbon dioxide at atmospheric pressure and recovered from the water upon de-carbonation of the mixture. Liquid chromatography-mass spectrometry (LC-MS) showed that the extracted lipids contained high concentrations of long chain tri-, di- and mono-acylglycerols, no phospholipids, and only 4-8% of residual solvent. Unlike extractions with conventional organic solvents, this new method requires neither distillation nor the use of volatile, flammable or chlorinated organic solvents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
290629
N,N-Dimethylcyclohexylamine, 99%
C8H17N