EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and antimicrobial activity of amide derivatives of polyether antibiotic-salinomycin.
PMID 22721714
Abstract
For the first time a direct and practical approach to the synthesis of eight amide derivatives of polyether antibiotic-salinomycin is described. The structure of allyl amide (3a) has been determined using X-ray diffraction. Salinomycin and its amide derivatives have been screened for their in vitro antimicrobial activity against the typical gram-positive cocci, gram-negative rods and yeast-like organisms, as well as against a series of clinical isolates of methicillin-resistant Staphylococcus aureus and methicillin-sensitive S. aureus. Amides of salinomycin have been found to show a wide range of activities, from inactive at 256 μg/mL to active with MIC of 2 μg/mL, comparable with salinomycin. As a result, phenyl amide (3b) was found to be the most active salinomycin derivative against gram-positive bacteria, MRSA and MSSA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S4526
Salinomycin, from Streptomyces albus, ≥98% (HPLC)
C42H70O11