EMAIL THIS PAGE TO A FRIEND
American journal of respiratory cell and molecular biology
Actin filament reorganization is a key step in lung inflammation induced by systemic inflammatory response syndrome.
PMID 22721831
Abstract
Acute lung injury (ALI) induced by systemic inflammatory response syndrome (SIRS) is characterized by deterioration in pulmonary function and leukocyte-associated lung inflammation. Actin fragment (F-actin) reorganization is required for leukocyte activation, adhesion, and transcription of inflammatory factors. We tested the hypothesis that F-actin plays a central role in SIRS-induced ALI. ALI was produced in a rat model with extracorporeal circulation. Cytochalasin B (CB) pretreatment to block F-actin reorganization improved oxygenation and reduced BAL inflammatory factors and pulmonary neutrophil sequestration, but did not reduce the adhesive molecules of blood leukocytes. We challenged blood neutrophils with TNF-α in vitro to explore the underlying mechanisms. Upon activation, neutrophils became polarized and formed a protrusive leading edge, with an aggregation of CD11b molecules. This effect could be blocked by CB, leading to reduced neutrophil adhesion. In addition, after LPS challenge, we observed F-actin reorganization and the up-regulation of inflammatory factors in pulmonary monocytes, which could also be blocked by CB pretreatment. F-actin reorganization initiates lung inflammation via increased blood neutrophil adhesion and migration, and by the production of inflammatory factors by pulmonary monocytes. Thus, blocking F-actin reorganization may potentially prevent and treat SIRS-induced ALI.
Related Materials