EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Mechanochemically induced disordered structures of vincamine: the different mediation of two cross-linked polymers.
PMID 22721852
Abstract
The aims of this research were to prepare highly bioavailable binary cogrounds (vincamine-AcDiSol(®) or PVP-Cl) by means of a mechanochemical process and to study the mediation of each polymer in the induction of physical transformations of the drug. From a set of fifteen cogrounds for each crosslinked polymer, two samples were selected in each group on the basis of the AUC of in vitro dissolution profiles with the help of a statistical comparison. The chosen samples were analysed by means of TEM, XRPD, Raman-spectroscopy/imaging, SSNMR, also including the study of (1)H spin-lattice relaxation times. The research encompassed in vivo oral absorption studies in rats, pharmacokinetic analysis and physical stability studies during 1 year. An intimate drug-polymer mixing was found in the coground samples with domain average dimensions smaller than 100 Å; this reflected in a remarkable enhancement of the in vitro and in vivo bioavailability. Different disordered states were detected in the coground samples as a function of cogrinding time and the type and amount of polymer used. Though both crosslinked polymers produced a remarkable enhancement of the oral bioavailability, coground systems based on AcDiSol(®) are preferable in terms of pharmacokinetic performance and physical stability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
194719
Vincamine, 98%
C21H26N2O3