EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
Effect of short-term proton pump inhibitor treatment and its discontinuation on chromogranin A in healthy subjects.
PMID 22723311
Abstract
Chromogranin A (CgA) is used as a generic tumor marker for neuroendocrine tumors. Proton pump inhibitors (PPI) are known to increase CgA, but it is not clear to what extent, and there is little information on how long PPI need to be discontinued before the effect of PPI has disappeared. Furthermore, is it not known whether this PPI effect is dependent on the CgA assay used. The aim of the study was to determine the effect of 7-d treatment with a PPI and its discontinuation on CgA in serum and plasma comparing four CgA assays. Seventeen healthy subjects took lansoprazole 30 mg at bedtime for 7 d, and blood samples for CgA were obtained at baseline, d 7 of PPI use, and 1, 2, 4, and 7 d after discontinuation of the PPI. In all samples, CgA was measured using the following assays: Alpco (serum and plasma), Cis-Bio (serum and plasma), DAKO, and Cis-Bio radioisotope assay. When using the same assay, CgA was higher in plasma than in serum. Treatment with a PPI for 1 wk resulted in a significant (about 2.5-fold) increase in CgA with significant interindividual variation. After discontinuation of PPI, serum CgA gradually declined, with a half-life of 4-5 d. Short-term PPI use results in a significant increase of CgA in serum and plasma, an effect that is largely independent of the assay used. PPI need to be discontinued for 2 wk to fully eliminate their effect on CgA. This effect of PPI needs to be considered when interpreting results of CgA measurements.