EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of tryptamine derivatives via a direct, one-pot reductive alkylation of indoles.
PMID 22724919
Abstract
An efficient, one-pot reductive alkylation of indoles with N-protected aminoethyl acetals in the presence of TES/TFA is reported. It represents the first general method for the direct synthesis of tryptamine derivatives from indoles and nitrogen-functionalized acetals. This convergent and versatile approach employs safe and inexpensive reagents, proceeds under mild conditions, and tolerates several functional groups. The new procedure was efficiently applied to a gram-scale synthesis of both luzindole, a reference MT2-selective melatonin receptor antagonist, and melatonin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
193747
Tryptamine, ≥97%
C10H12N2
246557
Tryptamine hydrochloride, 99%
C10H12N2 · HCl