EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Post-trapping derivatization of radical-derived EPR-silent adducts: application to free radical detection by HPLC/UV in chemical, biochemical, and biological systems and comparison with EPR spectroscopy.
PMID 22724922
Abstract
Free radicals are conventionally detected by electron paramagnetic resonance (EPR) spectroscopy after being trapped as spin adducts. Albeit this technique has demonstrated utmost efficacy in studying free radicals, its application to biological settings is intrinsically hampered by the inevitable bioreduction of radical-derived paramagnetic adducts. Herein, we describe a reliable technique to detect and quantify free radical metabolites, wherein reduced alkyl- and phenyl-5,5-dimethyl-1-pyrroline N-oxide (DMPO) adducts are converted into ultrastable N-naphthoate esters. To mimic the ubiquitous in vivo microenvironment, bioreductants, exogenous thiols, and sodium borohydride were studied. Nitroxyl reduction was confirmed using EPR and triphenyltetrazolium chloride. The formation of the N-naphthoyloxy derivatives was established by liquid chromatography/mass spectrometry (LC/MS). The derivatives were chromatographed using a binary eluent. HPLC and internal standards were synthesized using Grignard addition. The labeled DMPO adduct is (1) fluorescent, (2) stable as opposed to nitroxyl radical adducts, (3) biologically relevant, and (4) excellently chromatographed. Applications encompassed chemical, biochemical, and biological model systems generating C-centered radicals. Different levels of phenyl radicals produced in situ from whole blood were successfully determined. The method is readily applicable to the detection of hydroxyl radical. Analogously, DMPO, the spin trap, could be detected with extreme sensitivity suitable for in vivo applications. The developed method proved to be a viable alternative to EPR, where for the first time the reductive loss of paramagnetic signals of DMPO-trapped free radicals is transformed into fluorescence emission. We believe the proposed methodology could represent a valuable tool to probe free radical metabolites in vivo using DMPO, the least toxic spin trap.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92688
5,5-Dimethyl-1-pyrroline N-oxide, for ESR-spectroscopy
C6H11NO