EMAIL THIS PAGE TO A FRIEND
Investigational new drugs
Bisantrene in advanced, hormone-resistant carcinoma of the prostate: an Illinois Cancer Council phase II study.
PMID 2272771
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4563
Bisantrene dihydrochloride, ≥98% (HPLC)
C22H22N8 ·2HCl