EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
The vasodilator mechanisms of sodium metabisulfite on precontracted isolated aortic rings in rats: signal transduction pathways and ion channels.
PMID 22727858
Abstract
Sodium metabisulfite (SMB) is most commonly used as a food additives, however few study was performed on the vasodilator effect of SMB. In the present paper, the vasodilator effects of SMB and roles of Ca(2+) and K(+) channels as well as the cGMP pathway on isolated rat aortic rings were studied. The results show that: (1) SMB could relax isolated aortic rings precontracted by norepinephrine in a concentration-dependent manner. The maximal endothelium-dependent vasorelaxation was approximately 20% whereas that not depending on the presence of the endothelium was more than 90%. (2) The vasorelaxant effects induced by 50 or 200 μM SMB were partially inhibited by iberiotoxin, NS-2028 or l-NNA. The vasorelaxation of 1000 μM SMB was partially inhibited by nifedipine or glibenclamide. The SMB induced vasorelaxation was partially inhibited by tetraethylammonium. These results led to the conclusions that the vasorelaxation of SMB at low concentrations (<400 μM) was endothelium-dependent and mediated by the cGMP pathway and BK(Ca) channel, but at high concentrations (>500 μM) was endothelium-independent and mediated by K(ATP) channel and L-type Ca(2+) channel. The maximal allowable concentration from China and the acceptable daily intake level from WHO of SMB as a food additive should be revised.