EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Condensation reactions of 3-oxo-2-arylhydrazonopropanals with active methylene reagents: formation of 2-hydroxy- and 2-amino-6-substituted-5-arylazonicotinates and pyrido[3,2-c]cinnolines via 6π-electrocyclization reactions.
PMID 22728352
Abstract
3-Oxo-3-phenyl-2-(p-tolylhydrazono)propanal (1a) undergoes condensation with ethyl cyanoacetate in acetic acid in the presence of ammonium acetate to yield either 2-hydroxy-6-phenyl-5-p-tolylazonicotinic acid ethyl ester (6a) or 2-amino-6-phenyl-5-ptolyl-azonicotinic acid ethyl ester (8), depending on the reaction conditions. Similarly, other 3-oxo-3-aryl-2-arylhydrazonopropanals 1a,b condense with active methylene nitriles 2c,d to yield arylazonicotinates 6b,c. In contrast, 2-[(4-nitrophenyl)-hydrazono]-3-oxo-3-phenyl-propanal (1c) reacts with ethyl cyanoacetate to yield ethyl 6-(4-nitrophenyl)-2-oxo-2,6-dihydropyrido[3,2–c]cinnoline-3-carboxylate (11), via a novel 6π-electrocyclization pathway. Finally, 3-oxo-2-(phenylhydrazono)-3-p-tolylpropanal (1d) condenses with 2a-c to yield pyridazinones 13a-c.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W292303
Propionaldehyde, ≥97%, FG
C3H6O
538124
Propionaldehyde, reagent grade, 97%
C3H6O