EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Simple, rapid and reliable preparation of [¹¹C]-(+)-α-DTBZ of high quality for routine applications.
PMID 22728363
Abstract
[¹¹C]-(+)-α-DTBZ has been used as a marker of dopaminergic terminal densities in human striatum and expressed in islet beta cells in the pancreas. We aimed to establish a fully automated and simple procedure for the synthesis of [¹¹C]-(+)-α-DTBZ for routine applications. [¹¹C]-(+)-α-DTBZ was synthesized from a 9-hydroxy precursor in acetone and potassium hydroxide with [¹¹C]-methyl triflate and was purified by solid phase extraction using a Vac tC-18 cartridge. Radiochemical yields based on [¹¹C]-methyl triflate (corrected for decay) were 82.3% ± 3.6%, with a specific radioactivity of 60 GBq/μmol. Time elapsed was less than 20 min from end of bombardment to release of the product for quality control.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T2952
Tetrabenazine, ≥98% (HPLC), solid
C19H27NO3