EMAIL THIS PAGE TO A FRIEND
The American journal of sports medicine
Augmentation of tendon healing with butyric acid-impregnated sutures: biomechanical evaluation in a rabbit model.
PMID 22729622
Abstract
Butyric acid (BA) has been shown to be angiogenic and to enhance transcriptional activity in tissue. These properties of BA have the potential to augment biological healing of a repaired tendon. To evaluate this possibility both biomechanically and histologically in an animal tendon repair model. Controlled laboratory study. A rabbit Achilles tendon healing model was used to evaluate the biomechanical strength and histological properties at 6 and 12 weeks after repair. Unilateral tendon defects were created in the middle bundle of the Achilles tendon of each rabbit, which were repaired equivalently with either Ultrabraid BA-impregnated sutures or control Ultrabraid sutures. After 6 weeks, BA-impregnated suture repairs had a significantly increased (P < .0001) Young's modulus and ultimate tensile strength relative to the control suture repairs. At 12 weeks, no statistical difference was observed between these measures. The histological data at 6 weeks demonstrated significantly increased (P < .005) vessel density within 0.25 mm of the repair suture in the BA-impregnated group. There was also an associated 42% increase in the local number of myofibroblasts in the BA samples relative to the controls at this time. By 12 weeks, these differences were not observed. Tendons repaired with BA-impregnated sutures demonstrated improved biomechanical properties at 6 weeks relative to control sutures, suggesting a neoangiogenic mechanism of enhanced healing through an increased myofibroblast presence. These findings demonstrate that a relatively simple alteration of suture material may augment early tendon healing to create a stronger repair construct during this time.