EMAIL THIS PAGE TO A FRIEND
Molecular diversity
A novel and efficient one-pot four-component tandem approach for the synthesis of pyran derivatives.
PMID 22729829
Abstract
A series of new pyran derivatives are efficiently synthesized in a one-pot four-component tandem reaction via Suzuki coupling followed by a three-component reaction from readily available 4-bromobenzaldehyde, activated methylene compounds, and carbonyl compounds. Single crystal X-ray structure of the synthesized product is presented. The versatility and efficiency of the proposed methodology has been demonstrated in the synthesis of novel heterocyclic molecules.