EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Microbial butyrate and its role for barrier function in the gastrointestinal tract.
PMID 22731715
Abstract
Butyrate production in the large intestine and ruminant forestomach depends on bacterial butyryl-CoA/acetate-CoA transferase activity and is highest when fermentable fiber and nonstructural carbohydrates are balanced. Gastrointestinal epithelia seem to use butyrate and butyrate-induced endocrine signals to adapt proliferation, apoptosis, and differentiation to the growth of the bacterial community. Butyrate has a potential clinical application in the treatment of inflammatory bowel disease (IBD; ulcerative colitis). Via inhibited release of tumor necrosis factor α and interleukin 13 and inhibition of histone deacetylase, butyrate may contribute to the restoration of the tight junction barrier in IBD by affecting the expression of claudin-2, occludin, cingulin, and zonula occludens poteins (ZO-1, ZO-2). Further evaluation of the molecular events that link butyrate to an improved tight junction structure will allow for the elucidation of the cofactors affecting the reliability of butyrate as a clinical treatment tool.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19215
Butyric acid, analytical standard
C4H8O2