EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Gambogic acid activates AMP-activated protein kinase in mammalian cells.
PMID 22732400
Abstract
AMP-activated protein kinase (AMPK) plays a key role in maintaining intracellular and whole-body energy homeostasis. Activation of AMPK has been shown to ameliorate the symptoms of metabolic diseases, such as type 2 diabetes and obesity. Here we show that gambogic acid (GB), a known antitumor agent, activates AMPK by increasing the phosphorylation of AMPKα and its downstream substrate ACC in various cell lines. Further study revealed that GB stimulated AMPK activity independent of upstream kinases. Moreover, the AMPK inhibitor, compound C, has no effects on the GB-induced AMPK activation. We also found that GB promptly increased intracellular ROS level, and antioxidants attenuated the ROS production. Interestingly, only the thiol antioxidants significantly abolished GB-enhanced AMPK activation. In addition, analysis of binding and dissociation kinetics indicated that GB bound to the AMPKα subunit. Collectively, these results suggest that GB may be a novel direct activator of AMPK.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G8171
Gambogic acid, ≥95% (HPLC), powder
C38H44O8