EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
A new benzofuran derivative from Flemingia philippinensis Merr. et Rolfe.
PMID 22732880
Abstract
A new prenylated benzofuran derivative, named flemiphilippinone A, was isolated together with ten known flavonoids from the roots of Flemingia philippinensis. Flemiphilippinone A was identified as (2S,3aS)-5-(1-hydroxy-3-(4-methoxyphenyl)-propylidene)-2-(2-hydroxypropan-2-yl)-3a,7-bis(3-methylbut-2-en-1-yl)tetrahydrobenzofuran-4,6(2H,5H)-dione, and its structure was established by a combination of HR EIMS, ¹H-NMR, ¹³C-NMR, HMQC, HMBC and NOESY spectra data.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B8002
2,3-Benzofuran, 99%
C8H6O