EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Highly selective colorimetric sensing pyrophosphate in water by a NBD-phenoxo-bridged dinuclear Zn(II) complex.
PMID 22733118
Abstract
A novel NBD-phenoxo-bridged dinuclear Zn(II) complex is found to be an effective colorimetric sensor for pyrophosphate (PPi) in pure aqueous solution over a wide pH range. This sensor shows high binding affinity (K(a)≈ 3 × 10(8) M(-1)) and high selectivity for PPi, and can be also used to assay the activity of pyrophosphatase in real time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
433314
Pyrophosphoric acid, technical grade
H4O7P2