EMAIL THIS PAGE TO A FRIEND
European journal of immunology
The tyrosine kinase Abl is a component of macrophage podosomes and is required for podosome formation and function.
PMID 22733220
Abstract
Myeloid leukocytes form actin-based plasma membrane protrusions, called podosomes, that are implicated in myeloid cell recruitment into tissues and cell migration within the interstitium. In this study, we show that tyrosine kinases of the Abl family are present in podosomes formed by murine and human macrophages. Silencing of Abl expression in bone marrow-derived macrophages and monocyte-derived macrophages by siRNA or Abl enzymatic inhibition with imatinib resulted in the disassembly of macrophage podosomes and the reduction of their capacity to degrade an extracellular matrix and migrate through matrigel matrices and endothelial cell monolayers. Additionally, macrophages deficient in Src-family kinases, which cross-talk with Abl in regulating macrophage migration, also demonstrated podosome disassembly. These findings suggest that podosome disassembly induced by Abl targeting may inhibit podosome-dependent functions such as leukocyte recruitment into inflammatory sites and osteoclast-dependent bone resorption.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB1130 Anti-c-abl Antibody, clone 8E9, clone 8E9, Chemicon®, from mouse