EMAIL THIS PAGE TO A FRIEND
The Analyst
Development of a new and environment friendly hollow fiber-supported liquid phase microextraction using vesicular aggregate-based supramolecular solvent.
PMID 22733347
Abstract
Hollow fiber-based liquid phase microextraction (HF-LPME) using conventional solvents is limited by their relative instability and high volatility. The use of supramolecular solvents as a liquid membrane phase could overcome these inconveniences due to their negligible vapour pressure and high viscosity. In the present study, a novel and highly flexible method was developed based on supramolecular solvents constructed of vesicles of decanoic acid, which were used for the first time as a solvent in HF-LPME. This solvent is produced from the coacervation of decanoic acid aqueous vesicles by the action of tetrabutylammonium (Bu(4)N(+)). In this work, halogenated anilines as model compounds were extracted from water samples into a supramolecular solvent impregnated in the pores and also filled inside the porous polypropylene hollow fiber membrane. The extracted anilines were separated and determined by high-performance liquid chromatography. The technique requires minimal sample preparation time and toxic organic solvent consumption, and provides a significant advantage over conventional analytical methods. The important parameters influencing the extraction efficiency were studied and optimized utilizing two different optimization methods: one variable at a time and the Box-Behnken design. Under the optimum conditions, the preconcentration factors were in the range of 74 to 203. Linearity of the method was obtained in the range of 1.0-100 μg L(-1) with the correlation coefficients of determination (R(2)) ranging from 0.9901 to 0.9986. The limits of detection for the target anilines were 0.5-1.0 μg L(-1). The relative standard deviations varied from 3.9% to 6.0%. The relative recoveries of the three halogenated anilines from water samples at a spiking level of 20.0 μg L(-1) were in the range of 90.4-107.4%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21409
Decanoic acid, analytical standard
C10H20O2
C4151
Sodium decanoate, ≥98%
C10H19NaO2
651664
Tetrabutylammonium azide
C16H36N4
86868
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
C16H37NO4S
86853
Tetrabutylammonium bisulfate, for ion pair chromatography, LiChropur, ≥99.0%
C16H37NO4S
98479
Tetrabutylammonium bisulfate solution, for ion pair chromatography, LiChropur, concentrate, ampule
C16H37NO4S
72453
Tetrabutylammonium bisulfate solution, ~55% in H2O
C16H37NO4S
193119
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
C16H36BrN
426288
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
C16H36BrN
86857
Tetrabutylammonium bromide, for ion pair chromatography, LiChropur, ≥99.0%
C16H36BrN
462144
Tetrabutylammonium bromide solution, 50 wt. % in H2O
C16H36BrN
86870
Tetrabutylammonium chloride, ≥97.0% (NT)
C16H36ClN
86852
Tetrabutylammonium chloride, for ion pair chromatography, LiChropur, ≥99.0%
C16H36ClN
358665
Tetrabutylammonium cyanide, 95%
C17H36N2
79989
Tetrabutylammonium cyanide, technical, ≥80%
C17H36N2
216143
Tetrabutylammonium fluoride solution, 1.0 M in THF
C16H36FN
361399
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
C16H36FN
155837
Tetrabutylammonium hydrogensulfate, 97%
C16H37NO4S
86854
Tetrabutylammonium hydroxide solution, ~40% in water, for ion chromatography
C16H37NO
230189
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
C16H37NO
178780
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
C16H37NO
86880
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
C16H37NO
86863
Tetrabutylammonium hydroxide solution, 54.0-56.0% in H2O
C16H37NO
140775
Tetrabutylammonium iodide, reagent grade, 98%
C16H36IN
86890
Tetrabutylammonium iodide, ≥99.0% (AT)
C16H36IN
86903
Tetrabutylammonium iodide, for ion pair chromatography, LiChropur, ≥99.0%
C16H36IN
359173
Tetrabutylammonium nitrate, 97%
C16H36N2O3
86893
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
C16H36ClNO4
86885
Tetrabutylammonium perchlorate, ≥98.0% (T)
C16H36ClNO4
86833
Tetrabutylammonium phosphate monobasic, puriss., ≥99.0% (T)
C16H38NO4P