EMAIL THIS PAGE TO A FRIEND
Current heart failure reports
Mid-regional pro-adrenomedullin in acute heart failure: a better biomarker or just another biomarker?
PMID 22733501
Abstract
Circulating biomarkers have become increasingly important in diagnosing and risk-stratifying patients with heart failure (HF). While the natriuretic peptides have received much focus in the past decade, there is increasing interest in the role of other circulating biomarkers such as mid-regional proadrenomedullin (MR-proADM), a stable peptide of the precursor of adrenomedullin (ADM), responsible for volume regulation and electrolyte homeostasis. Increased levels of MR-proADM are associated with an increased risk of mortality and morbidity in patients with HF, independent of natriuretic peptides. MR-proADM outperforms all other established markers in the identification of patients at highest risk of death, particularly death within 30xa0days. The prognostic superiority has consistently been shown for various cardiovascular disease states, including acute heart failure. In this article, we discuss the potential role of MR-proADM in the syndrome of acute heart failure and its implication on prognosis and risk stratification.
Related Materials