EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Solid-phase extraction in the determination of gold, palladium, and platinum.
PMID 22733506
Abstract
A simple classification of various sorbents and solid-phase extraction procedures used for preconcentration of trace levels of Au, Pd, and Pt from different sample types is proposed in this review article. The large variety of available sorbents/procedures has been organized according to expected mechanisms of sorption process (complex formation; ion exchange; adsorption; ion-imprinted or molecularly imprinted polymers); according to the kind of monomeric units of the polymer matrix as well as on the basis of the kind of functional group responsible for main performance characteristics (selectivity, capacity) of the sorbent. Advantages of chemically modified sorbents, sulfur-containing sorbent extractants, and ion-imprinted polymers, together with rational pretreatment by means of microwave treatments, scaling down of enrichment, and quantification by means of flow and flow injection approaches are given. Preferred instrumental techniques for quantification of ppb levels of Au, Pd, and Pt in prepared concentrates/column eluates are multielement instrumental techniques: inductively coupled plasma optical emission spectrometry (ICP-OES), and inductively coupled plasma mass spectrometry (ICPMS). Excellent limits of detection at picogram levels of these analytes are provided by electrothermal atomic absorption spectrometry (ETAAS), generally in single-element mode and the neutron activation analysis (NAA), while X-ray fluorescence spectrometry and flame AAS are rarely applied because of lack of sensitivity at sub-ppm levels of Au, Pd, and Pt. Some problems of atomic spectrometric quantification techniques and their representative limits of detection are given. Recent applications to geological, industrial, pharmaceutical, biological, and other materials are tabulated. References have been selected mostly from the period 1995 to 2010.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
373192
Palladium, evaporation slug, diam. × L 0.9 cm × 1.2 cm, 99.95% trace metals basis
Pd
373206
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Pd
348678
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Pd
287474
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Pd
348643
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Pd
411450
Palladium, foil, thickness 0.05 mm, 99.9% trace metals basis
Pd
267120
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Pd
203998
Palladium, granular, 99.99% trace metals basis
Pd
203939
Palladium, powder, 99.999% trace metals basis
Pd
464651
Palladium, powder, <75 μm, 99.9% trace metals basis
Pd
326666
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Pd
346985
Palladium, rod, diam. 6.25 mm, ≥99.9% trace metals basis
Pd
267082
Palladium, sponge, 99.9% trace metals basis
Pd
267112
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Pd
348694
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Pd
643181
Palladium, extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support, Degussa type E105CA/W
Pd
686468
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Pd
GF46569831
Palladium, foil, 6mm disks, thickness 0.025mm, as rolled, 99.99+%
Pd
GF29399158
Palladium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.95%
Pd
GF21228825
Palladium, foil, 8mm disks, thickness 0.025mm, as rolled, 99.99+%
Pd
GF79862833
Palladium, wire reel, 0.05m, diameter 1.0mm, as drawn, 99.99+%
Pd
GF22660264
Palladium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.95%
Pd
GF00436733
Palladium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.99+%
Pd
GF85707513
Palladium, wire reel, 0.5m, diameter 1.0mm, hard, 99.95%
Pd
GF76572940
Palladium, wire reel, 1m, diameter 0.025mm, as drawn, 99.9%
Pd
GF96794409
Palladium, wire reel, 25m, diameter 0.125mm, hard, 99.95%
Pd
GF58966037
Palladium, wire reel, 2m, diameter 0.5mm, as drawn, 99.95%
Pd
GF24288790
Palladium, wire reel, 5m, diameter 0.25mm, hard, 99.95%
Pd
GF02178761
Palladium, wire reel, 2m, diameter 0.025mm, as drawn, 99.9%
Pd
GF93105547
Palladium, wire reel, 2m, diameter 0.02mm, as drawn, 99.95%
Pd