Luminescence : the journal of biological and chemical luminescence
Photoluminescence of Alq3 - and Tb-activated aluminium-tris(8-hydroxyquinoline) complex for blue chip-excited OLEDs.
PMID 22733610
The tris(8-hydroxyquinoline)-aluminium complex is the most important and widely studied as electron transporting and green light emitting material. Alq(3) and Tb(x) Al((1-x)) q(3) have been synthesized (where x = 0.1, 0.3, 0.5, 0.7 and 0.9) and blended films of Alq(3) and Tb(x) Al((1-x)) q(3) with PMMA and PS at different percentage weight (wt%) concentrations (e.g., 0.1, 1, 5, 10, 25 and 50 wt%) have been prepared. The synthesized materials and their blended thin films have been characterized by a photoluminescence (PL) technique; the synthesis and PL characterization are reported in this paper. The synthesized metal complex shows bright emission of green light with blue light excitation (440 nm) and the prepared Tb(x) Al((1-x)) q(3) phosphor may be applicable in blue chip-excited OLEDs for the newly developed wallpaper lighting technology.