EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Effects of penetration enhancers on curcumin transdermal drug delivery].
PMID 22734426
Abstract
To study the effects of penetration enhancers and their combinations on the curcumine transdermal drug delivery (CUR-TDDS). The penetration rate of curcumin through rat skin in vitro was measured using Valia-Chien diffusion cells, and orthogonal design method was set up for experimental design. The optimum penetration enhancers were: 3% hydroxypropyl beta cyclodextrins (HP-beta-CD), 9% borneol and 3% peppermint oil. The HP-beta-CD has the most potent enhancing effect.