EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Enantioselective conversion of primary alcohols to α-exo-methylene γ-butyrolactones via iridium-catalyzed C-C bond-forming transfer hydrogenation: 2-(alkoxycarbonyl)allylation.
PMID 22734694
Abstract
Upon exposure of acrylic ester 1 to alcohols 2a-i in the presence of a cyclometalated iridium catalyst modified by (-)-TMBTP, catalytic C-C coupling occurs, providing enantiomerically enriched 5-substituted α-exo-methylene γ-butyrolactones 3a-i. Bromination of the methylene butyrolactone products followed by zinc-mediated reductive aldehyde addition provides the disubstituted α-exo-methylene γ-butyrolactones 6a and 6b with good to excellent levels of diastereoselectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
226416
α-Methylene-γ-butyrolactone, 97%
C5H6O2