EMAIL THIS PAGE TO A FRIEND
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.]
A running modification of the percutaneous buried vertical mattress.
PMID 22734838
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
719846
Resomer® X 206 S, poly(dioxanone)