EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Lewis base mediated dismutation of trichlorosilane.
PMID 22735667
Abstract
An abnormal N-heterocyclic carbene (aNHC) has been used as a Lewis base to initiate dismutation of trichlorosilane. This report presents the reactivity differences of a normal N-heterocyclic carbene (NHC) versus aNHC with heavier group 14 elements. Three novel compounds (NHC)(2)·SiCl(2)H(2) (2), aNHC·SiCl(2)H(2) (3), and aNHC·GeCl(2) (4) have been synthesized and characterized by single crystal X-ray analysis, solid-state NMR and DFT calculations.