EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Study on chemical constituents of Drosera peltata var. multisepala].
PMID 22737855
Abstract
Chemical investigatation of Drosera peltata var. multisepala led to the isolation of eleven compounds using various chromatographic techniques. The structures of these compounds were elucidated as isoshinanolone-4-O-beta-D-glucoside (1), isoshinanolone (2), epi-isoshinanolone (3), plumbagin (4), droserone (5), droserone-5-O-glucoside (6), quercetin (7), kaempferol (8) , gossypetin-8-O-glucoside (9), 3,3'-dimethoxy ellagic acid (10), and ellagic acid (11) by their physicochemical properties and spectral data analysis. Compound 1 was a new compound. Compounds 3, 8, 10, and 11 were isolated from this plant for the first time.