EMAIL THIS PAGE TO A FRIEND
Zhongguo ying yong sheng li xue za zhi = Zhongguo yingyong shenglixue zazhi = Chinese journal of applied physiology
[Effect of polydatin on expression of cdk5 in the prefrontal cortex of rats with chronic alcoholism].
PMID 22737920
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
572691
3,4′-5-Trihydroxystilbene-3-β-D-glucopyranoside, 97%
C20H22O8
15721
Polydatin, ≥95% (HPLC)
C20H22O8