EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Uranium(III)-mediated C-C-coupling of terminal alkynes: formation of dinuclear uranium(IV) vinyl complexes.
PMID 22738030
Abstract
The previously reported uranium(III) complex [(((Ad)ArO)(3)N)U(III)(DME)] (1; Ad = adamantane, DME = 1,2-dimethoxyethane) reacts with the terminal bis-alkynes 1,7-octadiyne or 1,6-heptadiyne in C-C-coupling reactions to form the uranium(IV) vinyl complexes [{(((Ad)ArO)(3)N)U(IV)}(2)(μ-η(2):η(1)-1,2-(CH)(2)-cyclohexane)] (2) and [{(((Ad)ArO)(3)N)U(IV)}(2)(μ-η(2):η(2)-1,2-(CH)(2)-cyclopentane)] (3). With the monoalkynes 1-hexyne or 4-(t)butyl-phenylacetylene, the complexes [{(((Ad)ArO)(3)N)U(IV)}(2)(μ-η(2)(C1):η(1)(C4)-2-(n)Bu-1,3-octadiene)] (4) and [{(((Ad)ArO)(3)N)U(IV)}(2)(μ-η(2)(C4):η(1)(C1)-1,3-di-(p-(t)Bu-phenyl)butadiene))] (5), are formed. These are the first four examples of uranium vinyl complexes that are reported and crystallographically characterized. In addition, detailed DFT calculations are presented to establish a possible mechanism for their formation and explain the differences found for the coordination of the hydrocarbon fragments. In contrast to a previously proposed monometallic pathway for catalytic hydroamination of alkynes and alkyne dimerization involving a uranium vinyl intermediate at uranium(III) complexes, the calculations clearly support a bimetallic mechanism, since its transition states are energetically the most favored.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161292
1,7-Octadiyne, 98%
C8H10