EMAIL THIS PAGE TO A FRIEND
American journal of veterinary research
Pharmacokinetics of penciclovir in healthy cats following oral administration of famciclovir or intravenous infusion of penciclovir.
PMID 22738064
Abstract
To investigate the pharmacokinetics of penciclovir in healthy cats following oral administration of famciclovir or IV infusion of penciclovir. 6 cats. Cats received famciclovir (40 [n = 3] or 90 [3] mg/kg, PO, once) in a balanced crossover-design study; the alternate dose was administered after a ≥ 2-week washout period. After another washout period (≥ 4 weeks), cats received an IV infusion of penciclovir (10 mg/kg delivered over 1 hour). Plasma penciclovir concentrations were analyzed via liquid chromatography-mass spectrometry at fixed time points after drug administration. Mean ± SD maximum plasma concentration (C(max)) of penciclovir following oral administration of 40 and 90 mg of famciclovir/kg was 1.34 ± 0.33 μg/mL and 1.28 ± 0.42 μg/mL and occurred at 2.8 ± 1.8 hours and 3.0 ± 1.1 hours, respectively; penciclovir elimination half-life was 4.2 ± 0.6 hours and 4.8 ± 1.4 hours, respectively; and penciclovir bioavailability was 12.5 ± 3.0% and 7.0 ± 1.8%, respectively. Following IV infusion of penciclovir (10 mg/kg), mean ± SD penciclovir clearance, volume of distribution, and elimination half-life were 4.3 ± 0.8 mL/min/kg, 0.6 ± 0.1 L/kg, and 1.9 ± 0.4 hours, respectively. Penciclovir pharmacokinetics following oral administration of famciclovir were nonlinear within the dosage range studied, likely because of saturation of famciclovir metabolism. Oral administration of famciclovir at 40 or 90 mg/kg produced similar C(max) and time to C(max) values. Therefore, the lower dose may have similar antiviral efficacy to that proven for the higher dose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7932
Famciclovir, ≥98% (HPLC)
C14H19N5O4
P0035
Penciclovir
C10H15N5O3