EMAIL THIS PAGE TO A FRIEND
Medicina clinica
[Native valve endocarditis due to Staphylococcus aureus].
PMID 22743276
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9393
Cloxacillin sodium salt monohydrate, β-lactamase-resistant antibiotic
C19H17ClN3NaO5S · H2O