EMAIL THIS PAGE TO A FRIEND
Investigative Ophthalmology & Visual Science
Refractive lenticule re-implantation after myopic ReLEx: a feasibility study of stromal restoration after refractive surgery in a rabbit model.
PMID 22743323